Learn More
Abcam 560/585 nm Extracellular Vesicle Membrane Staining Kit, 100 TESTS
Description
- Assay type: Cell-based
- Detection method: Fluorescent
- Applications: Flow Cyt
- Sample types: Adherent cells, Suspension cells
- Reactive species: Mammals
- Results type: Quantitative
- Hands-on time: 30 seconds
- Labeling time: < 4 hours
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | Extracellular Vesicle Membrane |
| Reactivity | Mammals |
| Format | Kit |
| For Use With (Equipment) | Flow Cytometer |
| No. of Reactions | 100 Tests |
| Detection Method | Fluorescence |
| Content And Storage | Store at -20°C. |
| Cross Reactivity | Mammals |
| Sample Type | Adherent cells, Suspension cells |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.