Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Polyaniline (emeraldine salt) short chain, grafted to lignin

Catalog No. 56112610G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-112-610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-112-610G Supplier Sigma Aldrich Supplier No. 56112610G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Polyaniline (emeraldine salt) short chain, grafted to lignin (PANI-LIG) is an intrinsic conductive polymer with good electrical and optical properties. PANI is grafted with lignin to enhance the tensile strength and the bulk modulus of the polymer. It can be used for a variety of optoelectronic applications.PANI-LIG can be used in a variety of applications such as energy harvesting, dye sensitized solar cells, biosensors, and synthesis of ruthenium nanoparticles.Shake well before use.

Specifications

CAS 313949-90-5
MDL Number MFCD03458091
Quantity 10 g
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.