Learn More
Assay Technology 566 Organic Vapor Monitor | 4 chemicals | box of 5
SDP
Supplier:  Assay Technology X5664
Encompass_Preferred
The 566 Organic Vapor Monitor is our most versatile and popular organic vapor badge. The 566 utilizes our ChemDisk II configuration with a flexible, inert charcoal wafer, making it smaller and much easier to use than other badges on the market. The 76-hole sampling grid allows for sampling rates around 8 mL/min. While the 8-hour reporting limits vary from chemical to chemical (see our Sampling Guide), most are in the 0.1 ppm range, well below most OSHA PELs and NIOSH RELs. In addition, the 566 badge has much more sampling capacity than most other badge and tube methods.
Catalog No. NC1300310
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.