Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich tert-Butylsulfinyl chloride

Catalog No. 5694375G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-943-75G 5 g
56-943-71G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 56-943-75G Supplier Sigma Aldrich Supplier No. 5694375G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Tert-butylsulfinyl chloride can be synthesized by using tert-butylmagnesium chloride as the starting material via a multi-step synthesis process.tert-butylsulfinyl chloride can react with primary and secondary amines to form tert-butylsulfinamides, which on oxidation forms base stable tert-butylsulfonamide
  • It can also react with 1-thiosugars to form the corresponding carbohydrate thionolactones

Specifications

CAS 31562-43-3
Density 1.135 g/mL (at 25°C (literature))
Boiling Point 53°C to 54°C (19 mmHg)
Molecular Formula C4H9ClOS
Refractive Index n20/D 1.4830 (literature)
Linear Formula (CH3)3 CS(O)Cl
Quantity 5 g
MDL Number MFCD04974074
Molecular Weight (g/mol) 140.63
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.