Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Zinc Nanopowder 40-60 Nm Avg. Part. Size ≥99% Trace Metals Basis

Catalog No. 5780025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-800-25G 5 g
57-800-2250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 57-800-25G Supplier Sigma Aldrich Supplier No. 5780025G
Add to Cart
Add to Cart
This item is not returnable. View return policy

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Zinc Nanopowder 40-60 Nm Avg. Part. Size ≥99% Trace Metals Basis

Specifications

CAS 7440-66-6
Melting Point 420°C
Density 7.133 g/mL (at 25°C)
Boiling Point 907°C
Molecular Formula Zn
Linear Formula Zn
Quantity 5 g
MDL Number MFCD00011291
Molecular Weight (g/mol) 65.39
Percent Purity ≥99% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.