Learn More
Eppendorf™ 5804/5810 Series Four-Place Swinging Bucket Tube Adapters
Catalog No. 0540096 Shop All Eppendorf North America Products
Change view
Click to view available options
Bore Size (Metric):
1.1 cm
1.3 cm
1.6 cm
1.75 cm
3.1 cm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bore Size (Metric)
05-400-96 1.1 cm
05-400-98 1.6 cm
05-400-100 1.75 cm
05-400-103 3.1 cm
05-400-97 1.3 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 05-400-96 Supplier Eppendorf™ Supplier No. 022637525
Add to Cart
Add to Cart
Fit in buckets supplied with 05-400-94 swinging bucket rotor to accept tubes from 1.5 to 100mL
Adapters are sold in packs of two. Order the proper number of adapters to fully equip your rotor.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) A-4-44 swing bucket rotor for 5804/5804R/5810/5810R centrifuge
Bore Size (Metric) 1.1 cm