Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Di(propylene glycol) dibenzoate

Catalog No. 5972361L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-723-61L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 59-723-61L Supplier Sigma Aldrich Supplier No. 5972361L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Di(propylene glycol) dibenzoate (DPGDB) is a widely used plasticizer that has ether linkages linked with two benzoate groups.DPGDB may be used as a diluent for the preparation of polysulfone membranes by heat induced phase separation. It finds potential applications in water treatment and food processing. It may also be used as a plasticizer with poly(vinyl) chloride (PVC) for the fabrication of diamond coated PVC.

Specifications

CAS 27138-31-4
Density 1.12 g/mL (at 25°C)
Boiling Point 232°C (5 mmHg)
Molecular Formula C20H22O5
Refractive Index n20/D 1.528 (literature)
Linear Formula (C6H5CO2C3H6)2 O
Quantity 1 L
Molecular Weight (g/mol) 342.39
Percent Purity 75%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.