Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5alpha-Cholestan-3beta-ol, 98%

Catalog No. AAL0862406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0862406 5 g
AAL0862414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0862406 Supplier Thermo Scientific Chemicals Supplier No. L0862406
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 80-97-7 | C27H48O | 388.68 g/mol

5alpha-Cholestan-3beta-ol is used as a standard in lipid analysis using HPLC. It acts as a derivitized steroid compound. It is used in agrochemical, pharmaceutical and dyestuff field .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5alpha-Cholestan-3beta-ol is used as a standard in lipid analysis using HPLC. It acts as a derivitized steroid compound. It is used in agrochemical, pharmaceutical and dyestuff field .

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-97-7
Molecular Formula C27H48O
Molecular Weight (g/mol) 388.68
MDL Number MFCD00066413
InChI Key QYIXCDOBOSTCEI-UHFFFAOYNA-N
Synonym coprostan-3alpha-ol
PubChem CID 44135456
IUPAC Name (10S,13R)-10,13-dimethyl-17-(6-methylheptan-2-yl)-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES CC(C)CCCC(C)C1CCC2C3CCC4CC(O)CCC4(C)C3CCC12C

Specifications

Melting Point 140°C to 144°C
Quantity 5 g
Beilstein 2418594
Merck Index 14,2200
Solubility Information Insoluble in water.
Optical Rotation +24° (c=1 in Chloroform)
Formula Weight 388.68
Percent Purity 98%
Chemical Name or Material 5alpha-Cholestan-3beta-ol
EINECSNumber 201-315-8
RTECSNumber FZ6350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.