Learn More
Eppendorf 3P 5CUVETTE G1.0
Supplier:  Eppendorf 3P 6138000018
Encompass_Preferred
The Eppendorf µCuvette G1.0 microvolume measuring cell is a high-quality cuvette made of aluminum and quartz glass. It is the perfect tool for measuring high concentrations in small volumes. Higher sample concentrations require a shorter optical path length for measurements. With an optical path length of only 1 mm, the µCuvette G1.0 features a light path that is ten times shorter than the light path of standard cuvettes. This allows nucleic acid and protein concentrations to be measured with high reproducibility in a much higher concentration range without prior dilution. Due to the hydrophobic coating on the quartz glass only 1.5 µL nucleic acid or 3 µL protein sample are required for precise formation of the liquid column. Self-absorption of the µCuvette G1.0 is very low, meaning that the entire measuring range of the photometer can be used. • Microvolume measuring cell for measuring 1.5 – 10 µL sample volumes • Concentration determination of nucleic acids and proteins
Catalog No. 50-118-0054
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.