Learn More
Cytiva (Formerly Pall Lab) T-series Centramate™ Cassette, Omega PES membrane, 5 kDa molecular weight cut-off (MWCO), 0.1 m effective filtration area (EFA)
Catalog No. NC0733322 Shop All Cytiva Products
Encompass
Suitable for Process Development andSmall-Scale Production of 1 to 125 LFiltration area is easily expanded by adding additional membrane cassettesIdentical fluid path lengths provide precise linear scale-up to larger process systems available from Pall CorporationCassettes can be easily removed without disassembling system plumbingSystems are available with Type 316L stainless steel (Centramate system) or economically priced, extremely durable, ultra-high molecular weight polyethylene (Centramate PE system)Variety of membrane types provide versatility to the systems
Catalog No. NC0733322 Supplier Cytiva Supplier No. OS005T12
Add to Cart
Add to Cart
Suitable for Process Development andSmall-Scale Production of 1 to 125 LFiltration area is easily expanded by adding additional membrane cassettesIdentical fluid path lengths provide precise linear scale-up to larger process systems available from Pall CorporationCassettes can be easily removed without disassembling system plumbingSystems are available with Type 316L stainless steel (Centramate system) or economically priced, extremely durable, ultra-high molecular weight polyethylene (Centramate PE system)Variety of membrane types provide versatility to the systems