Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Grade 5M Sodium Chloride, Promega

Catalog No. PRV4221 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-V4221 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PR-V4221 Supplier Promega Corporation Supplier No. V4221
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Used in many molecular biology and forensic applications.

  • Quality tested and certified free of DNase, RNase and protease activity

Chemical Identifiers

CAS 7647-14-5
Molecular Formula ClNa
Molecular Weight (g/mol) 58.44
MDL Number MFCD00003477
InChI Key FAPWRFPIFSIZLT-UHFFFAOYSA-M
IUPAC Name sodium chloride
SMILES [Na+].[Cl-]

Specifications

Color Colorless
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Sodium Chloride
Recommended Storage 15 to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.