Learn More
Cayman Chemical FK-866, 658084-64-1, 5 mg
SDP
Supplier:  Cayman Chemical 132875
Encompass_Preferred
Molecular Formula C24H29N3O2, Purity ≥98%, Formula Weight 391.5, Synonyms: K 22.175; A highly specific non-competitive inhibitor of Nampt (Ki = 0.4 nM), causing gradual NAD+ depletion; directs delayed cell death by apoptosis in HepG2 cells (IC50 = ~1 nM).
Catalog No. NC0562856
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.