Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(5S)-5,6-Dihydro-6,6-dimethyl-5-phenyl-2H-1,4-oxazin-2-one 95.0+%, TCI America™
SDP

Catalog No. D32591G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D32591G Supplier TCI America Supplier No. D32591G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 723262-95-1
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.241
InChI Key AISCPSRYGCFAQP-NSHDSACASA-N
PubChem CID 11367698
IUPAC Name (3S)-2,2-dimethyl-3-phenyl-3H-1,4-oxazin-6-one
SMILES CC1(C(N=CC(=O)O1)C2=CC=CC=C2)C

Specifications

Melting Point 86°C
Color Yellow
Quantity 1 g
Formula Weight 203.24
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (5S)-5,6-Dihydro-6,6-dimethyl-5-phenyl-2H-1,4-oxazin-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.