Learn More
Chemglass Life Sciences Silicone Rubber Heater, 5” Wide x 30.62” Long for 290mm OD Vessels, Boot Latch Closure, 375 Watt, 110VAC, 6ft Cord with IEC Two Prong Plug
Supplier: Chemglass Life Sciences CLS1408290
Silicone rubber heater with boot latch, lace-style closure for 4 and 6 neck VSA (vertical side arm) bioreactors. Heaters are 375 watts and include a 6ft cord with an IEC two prong plug. Please reference the below chart for sizing the correct heater to your vessel. If you do not know the exact outside diameter the chart gives the approximate circumference which can be measured with a flexible tape measure.Item Number Vessel OD (mm) Approx. Circumference (mm) For Use with These 4 and 6 Neck VSA VesselsCLS-1408-238 238mm (9.37in) 747.7mm (29.4in) 3L, 6L, & 8LCLS-1408-290 290mm (11.4in) 911mm (35.9in) 15LCLS-1408-411 411mm (16.2in) 1921.2mm (50.8in) 36L
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.