Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5T4 Antibody (12B12C7C3) [Alexa Fluor 700] - Chimeric, R&D Systems™
SDP

Catalog No. NB590087
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB590087 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB590087 Supplier R&D Systems Supplier No. NBP345122AF700

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody

5T4 Monoclonal specifically detects 5T4 in Human samples. It is validated for ELISA, Flow Cytometry, Functional.

Specifications

Antigen 5T4
Applications ELISA, Flow Cytometry, Functional Assay
Classification Monoclonal
Clone 12B12C7C3
Conjugate Alexa Fluor 700
Dilution Flow Cytometry, ELISA, Functional
Formulation 50mM Sodium Borate
Gene Alias 5T4 oncofetal trophoblast glycoprotein, 5T4 oncotrophoblast glycoprotein, 5T4AG, 5T4-AG, 5T4M6P1,5T4 oncofetal antigen, trophoblast glycoprotein
Host Species Rabbit
Immunogen BALB/cAnNCrl mice or SJL/JorllcoCrl mice were immunized with either full length 5T4 or amino acid sequence 32-355 from 5T4 (Uniprot#: Q13641)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Immunology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 7162
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG κ
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.