Learn More
Tisch Scientific PTFE Syringe Filters, 5 um, 25mm, Luer-Lok/Luer Slip, Nonsterile, 100 per pack, SF17400
SDP
Supplier:  Tisch Scientific SF17400
Encompass_Preferred
Tisch PTFE Syringe filters are very versatile, proving to be effective for applications such as alkali filtration, strong acid solvent filtration, and organic solvent with strong chemical causticity filtration.
Catalog No. NC1407771
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.