Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Platinum™ II Green PCR Buffer (5X)

Catalog No. 14966123
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14966123 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14966123 Supplier Invitrogen™ Supplier No. 14966123
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

4 x 1.25 ml 5X Platinum II Green PCR buffer

Invitrogen 5X Platinum II Green PCR Buffer for fast and easy PCR

Invitrogen Platinum II Green PCR Buffer is an innovative buffer optimized for use with Platinum II Taq Hot-Start DNA Polymerase. Due to its unique composition, the annealing temperature is 60°C for most primer pairs designed following the general design rules. Isostabilizing molecules in the buffer increase primer–template duplex stability during the annealing step and contribute to enhanced specificity without the need to optimize annealing temperature for each primer pair.

Platinum II Green PCR Buffer provides 1.5 mM MgCl2 in the final reaction and is supplemented with a density reagent and two tracking dyes for direct loading of PCR products on gels.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5X
Content And Storage 5X Platinum II Green PCR buffer, 4 x 1.25 mL

Store at -20°C in a non-frost-free freezer.

Format Tube
Polymerase Platinum II Taq Hot-Start DNA Polymerase
Reaction Speed Fast
Product Type PCR Buffer
Quantity 5 mL
Volume 4 x 1.5 mL
Do the dyes in Platinum II Green PCR Buffer interfere with fragment separation of the PCR product on E-Gel agarose gels?

No. The tracing dyes (a blue and a yellow dye) in Platinum II Green PCR Buffer do not interfere with fragment separation on E-Gel agarose gels.

Do the dyes in Platinum II Green PCR Buffer interfere with PCR performance of Platinum II Taq Hot-Start DNA Polymerase?

No. The tracing dyes (a blue and a yellow dye) in Platinum II Green PCR Buffer do not interfere with PCR performance and do not change any enzyme features.

WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.