Learn More
Ace Glass, Inc. 5X5MM RASCHIG RINGS,PER LB
SDP
Supplier:  Ace Glass, Inc. 803304
Encompass_Preferred
Fabricated from Borosilicate glass. Wall weight on 5mm size is approximately 0.8.Sold per pound
Catalog No. NC2177658