Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-(1H-Pyrazol-1-yl)nicotinic acid, 97%

Catalog No. p-7032587
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH34104MD 250 mg
AAH3410403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH34104MD Supplier Thermo Scientific Chemicals Supplier No. H34104MD
Add to Cart

CAS: 253315-22-9 | C9H7N3O2 | 189.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 253315-22-9
Molecular Formula C9H7N3O2
Molecular Weight (g/mol) 189.17
MDL Number MFCD00140749
InChI Key QWFKXYLAKWFQLF-UHFFFAOYSA-N
Synonym 6-1h-pyrazol-1-yl nicotinic acid, 6-1h-pyrazol-1-yl pyridine-3-carboxylic acid, 6-pyrazol-1-yl-nicotinic acid, 6-pyrazol-1-yl pyridine-3-carboxylic acid, 6-pyrazolylpyridine-3-carboxylic acid, pyrazolylnicotinicacid, 6-pyrazol-1-ylnicotinic acid, 6-pyrazol-1-yl-nicotinicacid, 6-pyrazole-1-yl-nicotinic acid, 6-1h-pyrazol-1yl nicotinic acid
PubChem CID 2763614
SMILES OC(=O)C1=CN=C(C=C1)N1C=CC=N1

Specifications

Melting Point 272°C to 274°C
Quantity 250 mg
IUPAC Name 6-(1H-pyrazol-1-yl)pyridine-3-carboxylic acid
Formula Weight 189.2
Percent Purity 97%
Chemical Name or Material 6-(1H-Pyrazol-1-yl)nicotinic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.