Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-[2-Propyl-4-(4-pyridylazo)phenoxy]hexanoic Acid 98.0+%, TCI America™
SDP

Catalog No. P14361G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P14361G 1 g
P14365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P14361G Supplier TCI America Supplier No. P14361G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 312694-02-3
Molecular Formula C20H25N3O3
Molecular Weight (g/mol) 355.438
InChI Key RKSDFQDWVOJLAU-UHFFFAOYSA-N
PubChem CID 44629824
IUPAC Name 6-[2-propyl-4-(pyridin-4-yldiazenyl)phenoxy]hexanoic acid
SMILES CCCC1=C(C=CC(=C1)N=NC2=CC=NC=C2)OCCCCCC(=O)O

Specifications

Melting Point 173°C
Quantity 1 g
Formula Weight 355.44
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-[2-Propyl-4-(4-pyridylazo)phenoxy]hexanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.