Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-[(3-Chloropropyl)amino]-1,3-dimethyluracil 98.0+%, TCI America™
SDP

Catalog No. C29291G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C29291G Supplier TCI America Supplier No. C29291G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 34654-81-4
Molecular Formula C9H14ClN3O2
Molecular Weight (g/mol) 231.68
MDL Number MFCD00453355
InChI Key RPYBDDBZRQGARJ-UHFFFAOYSA-N
PubChem CID 3015699
IUPAC Name 6-(3-chloropropylamino)-1,3-dimethylpyrimidine-2,4-dione
SMILES CN1C(=CC(=O)N(C1=O)C)NCCCCl

Specifications

Melting Point 159°C
Color White-Yellow
Quantity 1 g
Formula Weight 231.68
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 6-[(3-Chloropropyl)amino]-1,3-dimethyluracil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.