Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 6-(4-Acetamido-1,8-Naphthalamido)Hexanoic Acid, Fluorescent Probe, >95% Purity, 10 mg

Catalog No. 502705359
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5359 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5359 Supplier Abcam Supplier No. AB14525210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fluorescent probe for labeling antisense oligonucleotides with >95% purity.

  • Molecular weight: 368.38 g/mol
  • Purity: >95%
  • Applications: Fluorescent probe for labeling antisense oligonucleotides
  • Storage: Store at -20°C
  • Shipping: Ambient - Can Ship with Ice
  • Composition: 100% 6-(4-Acetamido-1,8-naphthalamido)hexanoic acid

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Ambient
Target Molecule Antisense Oligonucleotides
Substrate Properties Chemical Substrate
Label or Dye Fluorescent Probe
Content And Storage Store at -20°C.
Format Vial
Detection Method Fluorescence
Buffer 100% 6-(4-Acetamido-1,8-naphthalamido)hexanoic acid
Quantity 10 mg
For Use With (Application) Labeling antisense oligonucleotides
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.