Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,6'-Dibromo-2,2'-bipyridyl 95.0+%, TCI America™
SDP

Catalog No. D39881G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39881G 1 g
D39885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39881G Supplier TCI America Supplier No. D39881G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 49669-22-9
Molecular Formula C10H6Br2N2
Molecular Weight (g/mol) 313.98
MDL Number MFCD00234587
InChI Key WZVWSOXTTOJQQQ-UHFFFAOYSA-N
PubChem CID 4407547
IUPAC Name 6,6'-dibromo-2,2'-bipyridine
SMILES BrC1=CC=CC(=N1)C1=NC(Br)=CC=C1

Specifications

Melting Point 225°C
Color White-Yellow
Quantity 1 g
Synonym 6,6′C-Dibromo-2,2′C-bipyridine
Formula Weight 313.98
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6,6′-Dibromo-2,2′-bipyridyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.