Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,6'-Dithiodinicotinic Acid 98.0+%, TCI America™
SDP

Catalog No. D256425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D256425G 25 g
D25645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D256425G Supplier TCI America Supplier No. D256425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15658-35-2
Molecular Formula C12H8N2O4S2
Molecular Weight (g/mol) 308.33
MDL Number MFCD00006454
InChI Key GSASOFRDSIKDSN-UHFFFAOYSA-N
PubChem CID 85040
IUPAC Name 6-[(5-carboxypyridin-2-yl)disulfanyl]pyridine-3-carboxylic acid
SMILES OC(=O)C1=CC=C(SSC2=CC=C(C=N2)C(O)=O)N=C1

Specifications

Color White-Yellow
Quantity 25 g
Synonym 6,6′-Dithiobis(3-pyridinecarboxylic Acid)
Formula Weight 308.33
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 6,6′-Dithiodinicotinic Acid
RTECSNumber US5684700
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.