Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,7-Diethoxy-1,2,3,4-tetrahydroisoquinoline Hydrochloride 98%, Thermo Scientific™

Catalog No. AC335740010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC335740010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC335740010 Supplier Thermo Scientific Chemicals Supplier No. 335740010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63905-65-7
Molecular Formula C13H20ClNO2
Molecular Weight (g/mol) 257.758
InChI Key GXDZORNOMJIKNT-UHFFFAOYSA-N
Synonym 6,7-diethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, 6,7-diethoxy-1,2,3,4-tetrahydroisoquinolinehydrochloride, isoquinoline,7-diethoxy-1,2,3,4-tetrahydro-, hydrochloride, isoquinoline,6,7-diethoxy-1,2,3,4-tetrahydro-, hydrochloride 1:1
PubChem CID 16232236
IUPAC Name 6,7-diethoxy-1,2,3,4-tetrahydroisoquinoline;hydrochloride
SMILES CCOC1=C(C=C2CNCCC2=C1)OCC.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Quantity 1 g
Formula Weight 257.76
Chemical Name or Material 6,7-Diethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.