Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,7-Dihydro-1H-indeno[5,4-b]furan-8(2H)-one 98.0+%, TCI America™
SDP

Catalog No. D50691G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50691G 1 g
D5069200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50691G Supplier TCI America Supplier No. D50691G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 196597-78-1
Molecular Formula C11H10O2
Molecular Weight (g/mol) 174.199
MDL Number MFCD09955085
InChI Key ZZUIZMWFNOKNLN-UHFFFAOYSA-N
Synonym 1,2,6,7-Tetrahydro-8H-indeno[5,4-b]furan-8-one
PubChem CID 11137616
IUPAC Name 1,2,6,7-tetrahydrocyclopenta[e][1]benzofuran-8-one
SMILES C1CC(=O)C2=C1C=CC3=C2CCO3

Specifications

Melting Point 135°C
Color White-Yellow
Quantity 1 g
Formula Weight 174.20
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6,7-Dihydro-1H-indeno[5,4-b]furan-8(2H)-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.