Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,7-Dimethoxyquinazoline-2,4-dione 98.0+%, TCI America™
SDP

Catalog No. D183925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D183925G 25 g
D18395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D183925G Supplier TCI America Supplier No. D183925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 28888-44-0
Molecular Formula C10H10N2O4
Molecular Weight (g/mol) 222.20
MDL Number MFCD00023889
InChI Key KWNQIIMVPSMYEM-UHFFFAOYSA-N
Synonym 6,7-dimethoxyquinazoline-2,4-dione, 2,4-dihydroxy-6,7-dimethoxyquinazoline, 6,7-dimethoxyquinazoline-2,4 1h,3h-dione, 6,7-dimethoxyquinazoline-2,4-diol, 6,7-dimethoxy-1,2,3,4-tetrahydroquinazoline-2,4-dione, 6,7-dimethoxy-1h,3h-2,4-quinazolindione, 2,4 1h,3h-quinazolinedione, 6,7-dimethoxy, 6,7-dimethoxy-2,4-quinazolinedione, 2,4-dihydroxy-6,7-dimethoxy quinazoline, 6,7-dimethoxy-2,4-dihydroxy quinazoline
PubChem CID 120081
IUPAC Name 6,7-dimethoxy-1,2,3,4-tetrahydroquinazoline-2,4-dione
SMILES COC1=C(OC)C=C2C(=O)NC(=O)NC2=C1

Specifications

Quantity 25 g
Formula Weight 222.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6,7-Dimethoxyquinazoline-2,4-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.