Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Acrylamidohexanoic Acid 98.0+%, TCI America™
SDP

Catalog No. A18961G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18961G 1 g
A18965G 5 g
A189625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A18961G Supplier TCI America Supplier No. A18961G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20766-85-2
Molecular Formula C9H15NO3
Molecular Weight (g/mol) 185.22
MDL Number MFCD06804452
InChI Key SAQWCPXBLNGTCC-UHFFFAOYSA-N
PubChem CID 324200
IUPAC Name 6-(prop-2-enamido)hexanoic acid
SMILES OC(=O)CCCCCNC(=O)C=C

Specifications

Melting Point 89°C
Color White-Yellow
Quantity 1 g
Formula Weight 185.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Acrylamidohexanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.