Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Amino-1,3-dimethyluracil 98.0+%, TCI America™
SDP

Catalog No. A1420500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1420500G 500 g
A142025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1420500G Supplier TCI America Supplier No. A1420500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6642-31-5
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.157
MDL Number MFCD00006552
InChI Key VFGRNTYELNYSKJ-UHFFFAOYSA-N
PubChem CID 81152
IUPAC Name 6-amino-1,3-dimethylpyrimidine-2,4-dione
SMILES CN1C(=CC(=O)N(C1=O)C)N

Specifications

Color Yellow
Quantity 500 g
Formula Weight 155.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Amino-1,3-dimethyluracil
EINECSNumber (5)-3381
RTECSNumber YQ8755000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.