Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Amino-2-mercaptobenzothiazole 97.0+%, TCI America™
SDP

Catalog No. A068225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A068225G 25 g
A06825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A068225G Supplier TCI America Supplier No. A068225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7442-07-1
Molecular Formula C7H6N2S2
Molecular Weight (g/mol) 182.26
MDL Number MFCD00059164
InChI Key IDPNFKLUBIKHSW-UHFFFAOYSA-N
PubChem CID 853584
IUPAC Name 6-amino-3H-1,3-benzothiazole-2-thione
SMILES C1=CC2=C(C=C1N)SC(=S)N2

Specifications

Quantity 25 g
Formula Weight 182.26
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Amino-2-mercaptobenzothiazole
RTECSNumber DL6488000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.