Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Amino-5-bromoquinoxaline 98.0+%, TCI America™
SDP

Catalog No. A291325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A291325G 25 g
A29135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A291325G Supplier TCI America Supplier No. A291325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50358-63-9
Molecular Formula C8H6BrN3
Molecular Weight (g/mol) 224.06
MDL Number MFCD00799024
InChI Key IRAOSCSPAYZRJE-UHFFFAOYSA-N
PubChem CID 14928442
IUPAC Name 5-bromoquinoxalin-6-amine
SMILES NC1=CC=C2N=CC=NC2=C1Br

Specifications

Melting Point 157°C
Quantity 25 g
Formula Weight 224.06
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Amino-5-bromoquinoxaline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.