Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Amino-7-bromo-9-methyl-7-deazapurine, 97%, Thermo Scientific Chemicals

Catalog No. AAH5989703
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5989703 1 g
AAH59897MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5989703 Supplier Thermo Scientific Chemicals Supplier No. H5989703
Add to Cart

It is used in the preparation of indoline derivatives as PERK inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the preparation of indoline derivatives as PERK inhibitors.

Solubility
Soluble in dichloromethane.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1337532-51-0
Molecular Formula C7H7BrN4
Molecular Weight (g/mol) 227.065
MDL Number MFCD20486185
InChI Key SCHJLAFNBDGWJN-UHFFFAOYSA-N
Synonym 5-bromo-7-methyl-7h-pyrrolo 2,3-d pyrimidin-4-amine, 5-bromo-7-methylpyrrolo 2,3-d pyrimidin-4-amine, 6-amino-7-bromo-9-methyl-7-deazapurine, 5-bromo-7-methyl-7h-pyrrolo-2,3-d pyrimidin-4-amine, 7h-pyrrolo 2,3-d pyrimidin-4-amine,5-bromo-7-methyl, 7h-pyrrolo 2,3-d pyrimidin-4-amine, 5-bromo-7-methyl
PubChem CID 66560938
IUPAC Name 5-bromo-7-methylpyrrolo[2,3-d]pyrimidin-4-amine
SMILES CN1C=C(C2=C1N=CN=C2N)Br

Specifications

Quantity 1 g
Beilstein 21981671
Solubility Information Soluble in dichloromethane.
Formula Weight 227.06
Percent Purity 97%
Chemical Name or Material 6-Amino-7-bromo-9-methyl-7-deazapurine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.