Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Aminofluorescein (isomer II) 95.0+%, TCI America™
SDP

Catalog No. A08641G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A08641G 1 g
A08645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A08641G Supplier TCI America Supplier No. A08641G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51649-83-3
Molecular Formula C20H13NO5
Molecular Weight (g/mol) 347.326
MDL Number MFCD00005051
InChI Key YOAWSYSKQHLFPM-UHFFFAOYSA-N
Synonym 6-aminofluorescein, fluoresceinamine isomer ii, 2-aminofluorescein, 6-amino-3',6'-dihydroxy-3h-spiro isobenzofuran-1,9'-xanthen-3-one, 6-amino-3',6'-dihydroxy-3h-spiro 2-benzofuran-1,9'-xanthen-3-one, 5-amino-3',6'-dihydroxyspiro 2-benzofuran-3,9'-xanthene-1-one, 6-amino-3',6'-dihydroxyspiro 2-benzofuran-1,9'-xanthene-3-one, 6-amino-3',6'-dihydroxyspiro isobenzofuran-1 3h ,9'-9h xanthene-3-one, spiro isobenzofuran-1 3h ,9'-9h xanthen-3-one, 6-amino-3',6'-dihydroxy
PubChem CID 103924
IUPAC Name 5-amino-3',6'-dihydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC2=C(C=C1N)C3(C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O)OC2=O

Specifications

Melting Point 285°C
Quantity 1 g
Formula Weight 347.33
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Aminofluorescein (isomer II)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.