Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Aminopenicillanic Acid, G-Biosciences
SCI_ED

Catalog No. S139774
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139774 10 g
S139775 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139774 Supplier G-Biosciences Supplier No. RC874
Add to Cart
Add to Cart

6-Aminopenicillanic acid (6-APA) is the central component of penicillin B-lactam antibiotics which are generated by Penicillium.

6-APA forms from penicillin G after being catalyzed by penicillin acylases. 6-APA is used as precursor for antibiotic compounds ampicillin and amoxicillin.

Chemical Identifiers

Molecular Weight (g/mol) 216.26 g/mol

Specifications

Boiling Point 460.2 ±45.0°C
Quantity 10 g
Percent Purity 98%
Grade Laboratory
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.