Learn More
Medchemexpress LLC 6-aminoquinolyl-N-hydroxysuccinimidyl carbamate | 148757-94-2 | MFCD01940654 | 285.25 g/mol | C14H11N3O4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY117695100MG
Encompass_Preferred
AQC (6-Aminoquinolyl-N-hydroxysuccinimidyl carbamate) is a fluorogenic derivatization reagent used to label primary and secondary amines, producing stable fluorescent derivatives for HPLC analysis with fluorescence detection. It enhances sensitivity for amino acid, peptide, and protein hydrolysate analysis.
  • Reacts rapidly with primary and secondary amines to form stable fluorescent derivatives.
  • Enables highly sensitive HPLC fluorescence detection of amino acids and peptides.
  • Ideal for pre-column derivatization workflows in amino acid and protein analysis.
  • Solid reagent; store sealed, away from moisture and light.
  • Compatible with common organic solvents; follow solvent storage recommendations.
Catalog No. 50-002-26438