Learn More
Sigma Organic Chemistry 6- Benzhydryloxy pyridi 1G
Supplier: Sigma Organic Chemistry OTV0006721G

6- Benzhydryloxy pyridi , 1G
About this item:
MDL #: MFCD11597302
Chemical Formula: C18H16N2O
Molecular Weight: 276.33
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.