Learn More
Apexbio Technology LLC AZD0156(Synonyms: AZD-0156, AZD0156 ATM inhibitor, ATM kinase inhibitor AZD0156, 1821428-35-6, ATM kinase inhibitor, AZD0156 inhibitor), 5mg, CAS: 1821428-35-6.
SDP
Supplier:  Apexbio Technology LLC B78225
Encompass_Preferred
AZD0156 (CAS number 1821428-35-6) is an orally bioavailable potent and selective small-molecule inhibitor of ataxia telangiectasia mutated (ATM) kinase a serine/threonine kinase within the phosphatidylinositol 3-kinase-related kinase (PIKK) family ATM kinase contributes critically to DNA damage response signaling activated by DNA double-strand breaks thus facilitating DNA repair checkpoint control genomic stability and cell fate decisions AZD0156 demonstrates sub-nanomolar inhibitory potency against cellular ATM signaling and exhibits over 1000-fold selectivity relative to other PIKK enzymes In preclinical models oral AZD0156 administration potentiates antitumor responses induced by double-strand break-generating agents Currently AZD0156 is undergoing early clinical trials assessing safety and preliminary efficacy for advanced cancers
Catalog No. 50-000-61228
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.