Learn More
Sigma Organic Chemistry 6-Bromo-2 7-dimethylimi 1G
Supplier: Sigma Organic Chemistry OTV0007491G

6-Bromo-2 7-dimethylimi , 1G
About this item:
MDL #: MFCD14546669
Chemical Formula: C9H9BrN2
Molecular Weight: 225.09
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.