Learn More
Medchemexpress LLC 6-bromo-2-hydroxy-3-methoxybenzaldehyde | 20035-41-0 | MFCD00046152 | 100.0% | 231.04 g·mol⁻¹ | C8H7BrO3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY107371100MG
Encompass_Preferred
6-Bromo-2-hydroxy-3-methoxybenzaldehyde is a brominated benzaldehyde derivative used as a research chemical and synthetic building block. It is a light yellow solid characterized for identity and purity and is applied in organic synthesis and biochemical studies requiring substituted benzaldehyde intermediates.
  • High purity suitable for analytical and synthetic work.
  • Light yellow solid for convenient handling and storage.
  • Compatible with common organic solvents and reaction conditions.
  • Characterized by LCMS and RP-HPLC for identity confirmation.
  • Useful as a building block in medicinal chemistry and probe synthesis.
Catalog No. 50-002-41278