Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromo-2-methylquinoline 98.0+%, TCI America™
SDP

Catalog No. B303925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B303925G 25 g
B30395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B303925G Supplier TCI America Supplier No. B303925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 877-42-9
Molecular Formula C10H8BrN
Molecular Weight (g/mol) 222.085
MDL Number MFCD00079724
InChI Key SQRYQSKJZVQJAY-UHFFFAOYSA-N
Synonym 6-bromoquinaldine, 2-methyl-6-bromoquinoline, 6-bromo-2-methyl-quinoline, quinoline, 6-bromo-2-methyl, 6-bromo-2-methyl-1-azanaphthalene, zlchem 926, pubchem5882, acmc-209qps, ksc494e7l
PubChem CID 522885
IUPAC Name 6-bromo-2-methylquinoline
SMILES CC1=NC2=C(C=C1)C=C(C=C2)Br

Specifications

Melting Point 104°C
Color Yellow
Quantity 25 g
Formula Weight 222.09
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromo-2-methylquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.