Learn More
Sigma Organic Chemistry 6-Bromo-3-iodo-1H-pyraz | 1G | MFCD22380432
Supplier: Sigma Organic Chemistry BLN000631G

6-Bromo-3-iodo-1H-pyraz , 1G
About this Item:
MDL #: MFCD22380432
Molecular Weight: 323.92
UNSPSC Code: 12352200
Chemical Formula: C6H3BrIN3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.