Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromohexanoyl chloride, 97%

Catalog No. p-7081193
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1106306 5 g
AAA1106314 25 g
AAA1106322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1106306 Supplier Thermo Scientific Chemicals Supplier No. A1106306
Only null left
Add to Cart

CAS: 22809-37-6 | C6H10BrClO | 213.499 g/mol

6-Bromohexanoyl chloride is used as intermediates, organic synthesis, chemical research, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromohexanoyl chloride is used as intermediates, organic synthesis, chemical research, agrochemicals and dyestuff.

Solubility
Reacts with water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed. Moisture Sensitive
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22809-37-6
Molecular Formula C6H10BrClO
Molecular Weight (g/mol) 213.499
MDL Number MFCD00000761
InChI Key HBPVGJGBRWIVSX-UHFFFAOYSA-N
Synonym hexanoyl chloride, 6-bromo, 6-bromohexanoic acid chloride, 6-bromohexanoylchloride, 6-bromocaproyl chloride, bromohexanoyl chloride, zlchem 435, acmc-209fyy, 6-bromo-hexanoylchloride, 6-bromo-hexanoyl chloride, 6-bromanylhexanoyl chloride
PubChem CID 89848
IUPAC Name 6-bromohexanoyl chloride
SMILES C(CCC(=O)Cl)CCBr

Specifications

Density 1.397
Boiling Point 119°C to 120°C (9 mmHg)
Flash Point >110°C (230°F)
Odor Acrid
Refractive Index 1.4875
Linear Formula Br(CH2)5COCl
Quantity 5 g
UN Number UN3265
Beilstein 1749739
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 213.5
Percent Purity 97%
Chemical Name or Material 6-Bromohexanoyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 245-236-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.