Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromoindazole 98.0+%, TCI America™
SDP

Catalog No. B40801G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40801G 1 g
B40805G 5 g
B408025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B40801G Supplier TCI America Supplier No. B40801G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 79762-54-2
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.04
MDL Number MFCD03265457
InChI Key WMKDUJVLNZANRN-UHFFFAOYSA-N
PubChem CID 17842471
IUPAC Name 6-bromo-1H-indazole
SMILES BrC1=CC=C2C=NNC2=C1

Specifications

Melting Point 183°C
Quantity 1 g
Synonym 6-bromoindazole, 6-bromo-2h-indazole, 1h-indazole, 6-bromo, 1h-indazole,6-bromo, pubchem7831, zlchem 1237, 6-bromo-1-h-indazole, 6-bromanyl-1h-indazole, acmc-209pi5
Formula Weight 197.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromoindazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.