Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromopyridine-2-carboxaldehyde, 97%

Catalog No. AAL1951806
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1951806 5 g
AAL1951803 1 g
AAL1951814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1951806 Supplier Thermo Scientific Chemicals Supplier No. L1951806
Add to Cart

CAS: 34160-40-2 | C6H4BrNO | 186.008 g/mol

6-Bromopyridine-2-carboxaldehyde is used in the study of a rhodium-catalyzed, reductive aldol coupling with divinyl ketones leading to syn ß-hydroxyenones. It is also used as a building block for Tris[(pyridyl)methyl]amine ligands.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromopyridine-2-carboxaldehyde is used in the study of a rhodium-catalyzed, reductive aldol coupling with divinyl ketones leading to syn ß-hydroxyenones. It is also used as a building block for Tris[(pyridyl)methyl]amine ligands.

Solubility
Soluble in dichloromethane, ether, ethyl acetate and methanol. Insoluble in water.

Notes
Store under inert gas. It is sensitive to air. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34160-40-2
Molecular Formula C6H4BrNO
Molecular Weight (g/mol) 186.008
MDL Number MFCD02683546
InChI Key QWFHFNGMCPMOCD-UHFFFAOYSA-N
Synonym 6-bromo-2-pyridinecarboxaldehyde, 6-bromopicolinaldehyde, 6-bromo-2-formylpyridine, 6-bromopyridine-2-carboxaldehyde, 2-bromo-6-formylpyridine, 6-bromo-pyridine-2-carbaldehyde, 2-pyridinecarboxaldehyde, 6-bromo, 2-bromopyridine-6-carboxaldehyde, 6-bromo-2-pyridine carboxaldehyde, 6-bromopicolylaldehyde
PubChem CID 2757009
IUPAC Name 6-bromopyridine-2-carbaldehyde
SMILES C1=CC(=NC(=C1)Br)C=O

Specifications

Melting Point 79°C to 83°C
Quantity 5 g
Beilstein 1524300
Sensitivity Air Sensitive
Solubility Information Soluble in dichloromethane,ether,ethyl acetate and methanol. Insoluble in water.
Formula Weight 186.01
Percent Purity 97%
Chemical Name or Material 6-Bromopyridine-2-carboxaldehyde
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.