Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromoquinoxaline 98.0+%, TCI America™
SDP

Catalog No. B44761G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44761G 1 g
B44765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44761G Supplier TCI America Supplier No. B44761G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50998-17-9
Molecular Formula C8H5BrN2
Molecular Weight (g/mol) 209.05
MDL Number MFCD00837757
InChI Key NOYFLUFQGFNMRB-UHFFFAOYSA-N
Synonym 6-bromo-quinoxaline, quinoxaline, 6-bromo, 6-bromobenzopyrazine, acmc-209krf, 6-bromoquinoxaline, ksc269e9f
PubChem CID 610939
IUPAC Name 6-bromoquinoxaline
SMILES BrC1=CC=C2N=CC=NC2=C1

Specifications

Melting Point 56°C
Boiling Point 149°C
Quantity 1 g
Formula Weight 209.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromoquinoxaline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.