Learn More
Abcam 6-Carboxyfluorescein N-hydroxysuccinimide Ester, Fluorescent Labeling Reagent, Solid, 10 mg
Description
- Molecular Weight: 473.39 g/mol
- Form: Solid
- Applications: Fluorescent labeling of peptides, proteins and nucleotides
- Composition: 100% 6-Carboxyfluorescein-N-hydroxysuccinimide ester
- Chemical Properties: Amine-reactive succinimidyl ester carboxyfluorescein; yields hydrolysis-resistant carboxyamides
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Peptides, Proteins, Nucleotides |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Label or Dye | 6-Carboxyfluorescein |
| Kit Contents | 10 mg 6-Carboxyfluorescein-N-hydroxysuccinimide ester |
| Form | Solid |
| Product Type | Protein Labeling Reagent |
| Detection Method | Fluorescence |
| Content And Storage | Store at ambient temperature. |
| Quantity | 10 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.