Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam 6-Carboxyfluorescein N-hydroxysuccinimide Ester, Fluorescent Labeling Reagent, Solid, 100 mg
Description
- Molecular weight: 473.39 g/mol
- Form: Solid
- Applications: Fluorescent labeling of peptides, proteins and nucleotides
- Features: Amine-reactive succinimidyl ester carboxyfluorescein which yields carboxyamides more resistant to hydrolysis than FITC conjugates
- Storage: Store at Ambient
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Label or Dye | 6-Carboxyfluorescein |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Target Molecule | Peptides, Proteins, Nucleotides |
| Form | Solid |
| Detection Method | Fluorescence |
| Product Type | Protein Labeling Reagent |
| Buffer | 100% 6-Carboxyfluorescein-N-hydroxysuccinimide ester |
| Content And Storage | Store at ambient temperature. |
| Quantity | 100 mg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?