Learn More
V&P Scientific Inc ASPIRATION MANIFOLD, 6 Channel, 24 Well, Deep Well, 19 Gauge, SS Tubes, 49mm Long, 20mm Spacing, Polycarbonate
Supplier:  V&P Scientific Inc VP 182C
Encompass
ASPIRATION MANIFOLD, 6 Channel, 24 Well, Deep Well Plate, 19 Gauge, Stainless Steel Tubes, 49mm Long, 20mm Center Spacing, Polycarbonate, for Costar #3524 Plates
Catalog No. NC9923908