Learn More
Sigma Organic Chemistry 6-chloro-2-naphthY sulf | 250mg | MFCD04037080
Supplier: Sigma Organic Chemistry CDS003604250MG

6-chloro-2-naphthY sulf , 250mg
About this Item:
MDL #: MFCD04037080
Molecular Weight: 261.12
UNSPSC Code: 12352200
Chemical Formula: C10H6Cl2O2S
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.