Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloro-3-formylchromone 98.0+%, TCI America™
SDP

Catalog No. C286925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C286925G 25 g
C28695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C286925G Supplier TCI America Supplier No. C286925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 42248-31-7
Molecular Formula C10H5ClO3
Molecular Weight (g/mol) 208.597
MDL Number MFCD00139138
InChI Key LIGLNCRTMDRUAO-UHFFFAOYSA-N
Synonym 6-chloro-3-formylchromone, 6-chloro-4-oxo-4h-chromene-3-carbaldehyde, chembl84898, 4h-1-benzopyran-3-carboxaldehyde,6-chloro-4-oxo, 6-chloro-4-oxo-4h-1-benzopyran-3-carboxaldehyde, zlchem 688, 3-formyl-6-chlorochromone, 6-chlorochromone-3-carboxaldehyde, 6-chloro-4-oxo-chromene-3-carbaldehyde, 6-chloro-4-oxo-1-benzopyran-3-carboxaldehyde
PubChem CID 463777
IUPAC Name 6-chloro-4-oxochromene-3-carbaldehyde
SMILES C1=CC2=C(C=C1Cl)C(=O)C(=CO2)C=O

Specifications

Melting Point 167°C
Color Yellow
Quantity 25 g
Formula Weight 208.60
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloro-3-formylchromone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.